Assay Details
Target Gene Details
Entrez Gene ID: | 445 |
Gene Name: | argininosuccinate synthase 1 |
Gene Aliases: |
ASS, CTLN1 |
Location: |
Chr.9:130444707-130501274 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASS1 | NM_000050.4 | NP_000041.2 | ||
| NM_054012.3 | NP_446464.1 | |||
| XM_005272200.3 | XP_005272257.1 | |||
| XM_017014729.1 | XP_016870218.1 | |||
| AK027126.1 | ||||
| AK293114.1 | ||||
| AK304625.1 | ||||
| AK315093.1 | ||||
| AW672752.1 | ||||
| BC009243.2 | AAH09243.1 | |||
| BC021676.1 | AAH21676.1 | |||
| BE207182.1 | ||||
| BF305325.1 | ||||
| BI603075.1 | ||||
| BX403885.2 | ||||
| DB496935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466597 | Chr.9:130443364 - 130460028 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73657381] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map