Assay Details
Target Gene Details
Entrez Gene ID: | 10541 |
Gene Name: | acidic nuclear phosphoprotein 32 family member B |
Gene Aliases: |
APRIL, PHAPI2, SSP29 |
Location: |
Chr.9:97983207-98015943 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANP32B | NM_006401.2 | NP_006392.1 | ||
| AK223454.1 | BAD97174.1 | |||
| AK313733.1 | ||||
| BC013003.2 | AAH13003.1 | |||
| BC019658.1 | AAH19658.1 | |||
| U70439.1 | AAB37579.1 | |||
| Y07569.1 | CAA68855.1 | |||
| Y07570.1 | CAA68856.1 | |||
| Y07969.1 | CAA69265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2540773 | Chr.9:97998181 - 97999833 on Build GRCh38 | Deletion |
|
| esv7209 | Chr.9:97998598 - 97999375 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map