Assay Details
Target Gene Details
Entrez Gene ID: | 56254 |
Gene Name: | ring finger protein 20 |
Gene Aliases: |
BRE1, BRE1A, hBRE1 |
Location: |
Chr.9:101533849-101563344 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF20 | NM_019592.6 | NP_062538.5 | ||
| XM_011518862.1 | XP_011517164.1 | |||
| AF265230.1 | AAK58539.1 | |||
| AK022532.1 | BAB14081.1 | |||
| AK314401.1 | ||||
| BC016691.1 | ||||
| BC063115.1 | AAH63115.1 | |||
| BC110584.1 | AAI10585.1 | |||
| BC110585.1 | AAI10586.1 | |||
| BC152309.1 | ||||
| DB273543.1 |
More Information
Set Membership: |
|

Genomic Map