Assay Details
Target Gene Details
Entrez Gene ID: | 100506422 |
Gene Name: | putative deoxyuridine 5'-triphosphate nucleotidohydrolase-like protein FLJ16323 |
Gene Aliases: |
- |
Location: |
Chr.9:26066675-26126108 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506422 | NM_001004352.2 | NP_001004352.1 | ||
| XM_017014175.1 | XP_016869664.1 | |||
| AK131322.1 | BAD18483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv613923 | Chr.9:26005489 - 26112750 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map