Assay Details
Target Gene Details
Entrez Gene ID: | 11168 |
Gene Name: | PC4 and SFRS1 interacting protein 1 |
Gene Aliases: |
DFS70, LEDGF, PAIP, PSIP2, p52, p75 |
Location: |
Chr.9:15464066-15511019 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSIP1 | NM_001128217.2 | NP_001121689.1 | ||
| NM_001317898.1 | NP_001304827.1 | |||
| NM_001317900.1 | NP_001304829.1 | |||
| NM_021144.3 | NP_066967.3 | |||
| NM_033222.4 | NP_150091.2 | |||
| AF063020.1 | AAC25167.1 | |||
| AF098482.1 | AAC97945.1 | |||
| AF098483.1 | AAC97946.1 | |||
| AF432220.1 | AAL99926.1 | |||
| AK096405.1 | 1 | 880 | ||
| BC044568.1 | AAH44568.2 | |||
| BC064135.1 | AAH64135.1 | |||
| BX649155.1 | 1 | 1025 | ||
| U94319.1 | AAB52589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517975 | Chr.9:15286448 - 15534072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map