Assay Details
Target Gene Details
Entrez Gene ID: | 6850 |
Gene Name: | spleen associated tyrosine kinase |
Gene Aliases: |
p72-Syk |
Location: |
Chr.9:90801680-90898560 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYK | NM_001135052.3 | NP_001128524.1 | ||
| NM_001174168.2 | NP_001167639.1 | |||
| NM_003177.6 | NP_003168.2 | |||
| XM_005252147.3 | XP_005252204.1 | |||
| XM_011518946.2 | XP_011517248.1 | |||
| AK075020.1 | ||||
| AK290927.1 | ||||
| BC001645.2 | AAH01645.1 | |||
| BC002962.1 | AAH02962.1 | |||
| BC011399.1 | AAH11399.1 | |||
| BM019270.1 | ||||
| DA587403.1 | ||||
| DB458719.1 | ||||
| L28824.1 | AAA36526.1 | |||
| Z29630.1 | CAA82737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621052 | Chr.9:90793616 - 90813635 on Build GRCh38 | Gain |
|
| nsv831652 | Chr.9:90765129 - 90934641 on Build GRCh38 | Gain |
|
| esv3621048 | Chr.9:90655588 - 91365881 on Build GRCh38 | Gain |
|
| nsv1038685 | Chr.9:90793892 - 90923447 on Build GRCh38 | Gain |
|
| esv3621051 | Chr.9:90788855 - 90818333 on Build GRCh38 | Gain |
|
| nsv1046989 | Chr.9:90591705 - 91038134 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79216110] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map