Assay Details
Target Gene Details
Entrez Gene ID: | 23424 |
Gene Name: | tudor domain containing 7 |
Gene Aliases: |
CATC4, PCTAIRE2BP, TRAP |
Location: |
Chr.9:97412020-97496125 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRD7 | NM_001302884.1 | NP_001289813.1 | ||
| NM_014290.2 | NP_055105.2 | |||
| XM_006717026.3 | XP_006717089.1 | |||
| AK294488.1 | ||||
| AK314853.1 | ||||
| BC028694.1 | AAH28694.1 | |||
| DB455041.1 |
More Information
Set Membership: |
|

Genomic Map