Assay Details
Target Gene Details
Entrez Gene ID: | 10655 |
Gene Name: | doublesex and mab-3 related transcription factor 2 |
Gene Aliases: |
- |
Location: |
Chr.9:1003237-1057554 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMRT2 | XM_011517690.2 | XP_011515992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891516 | Chr.9:892241 - 1158171 on Build GRCh38 | Gain |
|
| nsv1025752 | Chr.9:1003311 - 1124524 on Build GRCh38 | Gain |
|
| nsv613037 | Chr.9:643453 - 1138368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map