Assay Details
Target Gene Details
Entrez Gene ID: | 101926900 |
Gene Name: | DNAJB5 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.9:34985410-34989379 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB5-AS1 | NR_109756.1 | 2 | 429 | |
| AI198641.1 | 1 | 335 | ||
| DA679664.1 | 2 | 431 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6527 | Chr.9:34984490 - 35015049 on Build GRCh38 | Insertion |
|

Genomic Map