Assay Details
Target Gene Details
Entrez Gene ID: | 100499484 |
Gene Name: | SUGT1-1300002K09Rik pseudogene |
Gene Aliases: |
- |
Location: |
Chr.9:97238426-97297314 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100499484 | NR_036526.1 | |||
| AK301827.1 |
Target Gene Details
Entrez Gene ID: | 57653 |
Gene Name: | LOC100499484-C9orf174 readthrough |
Gene Aliases: |
- |
Location: |
Chr.9:97238426-97377295 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100499484-C9ORF174 | NR_036527.1 | |||
| NR_036528.1 | ||||
| NR_036529.1 |

Genomic Map