Assay Details
Target Gene Details
Entrez Gene ID: | 7111 |
Gene Name: | tropomodulin 1 |
Gene Aliases: |
D9S57E, ETMOD, TMOD |
Location: |
Chr.9:97501180-97601743 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMOD1 | NM_001166116.1 | NP_001159588.1 | ||
| NM_003275.3 | NP_003266.1 | |||
| AK314533.1 | ||||
| BC002660.2 | AAH02660.1 | |||
| DA015088.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113601530] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map