Assay Details
Target Gene Details
Entrez Gene ID: | 840 |
Gene Name: | caspase 7 |
Gene Aliases: |
CASP-7, CMH-1, ICE-LAP3, LICE2, MCH3 |
Location: |
Chr.10:113679162-113730909 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624603 | Chr.10:113581857 - 113745416 on Build GRCh38 | Gain |
|
| nsv1048819 | Chr.10:113499347 - 113757619 on Build GRCh38 | Gain |
|
| esv3624607 | Chr.10:113679337 - 113732388 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map