Assay Details
Target Gene Details
Entrez Gene ID: | 24138 |
Gene Name: | interferon induced protein with tetratricopeptide repeats 5 |
Gene Aliases: |
ISG58, P58, RI58 |
Location: |
Chr.10:89414568-89421002 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFIT5 | NM_012420.2 | NP_036552.1 | ||
| AK293346.1 | ||||
| AK312358.1 | ||||
| BC025786.1 | AAH25786.1 | |||
| DC392439.1 | ||||
| U34605.1 | AAA84934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819719 | Chr.10:89406006 - 89414843 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map