Assay Details
Target Gene Details
Entrez Gene ID: | 8030 |
Gene Name: | coiled-coil domain containing 6 |
Gene Aliases: |
D10S170, H4, PTC, TPC, TST1 |
Location: |
Chr.10:59788747-59906656 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC6 | NM_005436.4 | NP_005427.2 | ||
| BM672399.1 |
More Information
Set Membership: |
|

Genomic Map