Assay Details
Target Gene Details
Entrez Gene ID: | 5408 |
Gene Name: | pancreatic lipase related protein 2 (gene/pseudogene) |
Gene Aliases: |
PLRP2 |
Location: |
Chr.10:116620953-116645143 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNLIPRP2 | NM_005396.4 | NP_005387.2 | ||
| NR_103727.1 | ||||
| XM_017016339.1 | XP_016871828.1 | |||
| AK291492.1 | ||||
| AK308508.1 | ||||
| AK308511.1 | ||||
| AK308521.1 | ||||
| BC005989.2 | AAH05989.1 | |||
| CR456949.1 | CAG33230.1 | |||
| M93284.1 | AAA59533.1 |
More Information
Set Membership: |
|

Genomic Map