Assay Details
Target Gene Details
Entrez Gene ID: | 100528007 |
Gene Name: | BORCS7-ASMT readthrough (NMD candidate) |
Gene Aliases: |
C10orf32-AS3MT, C10orf32-ASMT |
Location: |
Chr.10:102854210-102901899 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS7-ASMT | NR_037644.1 |

Genomic Map