Assay Details
Target Gene Details
Entrez Gene ID: | 282973 |
Gene Name: | Janus kinase and microtubule interacting protein 3 |
Gene Aliases: |
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5 |
Location: |
Chr.10:132036068-132184852 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAKMIP3 | XM_011539676.2 | 1 | 169 | XP_011537978.1 |
| XM_017016096.1 | 1 | 168 | XP_016871585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain |
|
| esv3624956 | Chr.10:131249138 - 132223150 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| nsv7625 | Chr.10:132007295 - 132041165 on Build GRCh38 | Insertion |
|
| esv3624980 | Chr.10:131991638 - 132087199 on Build GRCh38 | Gain |
|
| nsv825650 | Chr.10:131914064 - 132056715 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs201255403] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map