Assay Details
Target Gene Details
Entrez Gene ID: | 56243 |
Gene Name: | KIAA1217 |
Gene Aliases: |
SKT |
Location: |
Chr.10:23694746-24547848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1217 | NM_001098500.2 | NP_001091970.1 | ||
| XM_017016429.1 | XP_016871918.1 | |||
| AB033043.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs77644717] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map