Assay Details
Target Gene Details
Entrez Gene ID: | 22978 |
Gene Name: | 5'-nucleotidase, cytosolic II |
Gene Aliases: |
GMP, NT5B, PNT5, SPG45, SPG65, cN-II |
Location: |
Chr.10:103088017-103193306 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
More Information
Set Membership: |
|

Genomic Map