Assay Details
Target Gene Details
Entrez Gene ID: | 84896 |
Gene Name: | ATPase family, AAA domain containing 1 |
Gene Aliases: |
AFDC1, FNP001, THORASE |
Location: |
Chr.10:87751512-87841345 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATAD1 | NM_001321967.1 | 10 | 3917 | NP_001308896.1 |
| NM_001321968.1 | 9 | 3827 | NP_001308897.1 | |
| NM_001321969.1 | 11 | 4007 | NP_001308898.1 | |
| NM_032810.3 | 10 | 4289 | NP_116199.2 | |
| NR_135914.1 | 11 | 4004 | ||
| XM_005270252.4 | 10 | 4011 | XP_005270309.1 | |
| XM_011540302.1 | 8 | 3682 | XP_011538604.1 | |
| XM_017016847.1 | 10 | 4158 | XP_016872336.1 | |
| XM_017016848.1 | 9 | 4161 | XP_016872337.1 | |
| XM_017016849.1 | 11 | 4130 | XP_016872338.1 | |
| XM_017016850.1 | 11 | 4181 | XP_016872339.1 |
More Information
Set Membership: |
|

Genomic Map