Assay Details
Target Gene Details
Entrez Gene ID: | 10529 |
Gene Name: | nebulette |
Gene Aliases: |
LASP2, LNEBL |
Location: |
Chr.10:20779973-21174187 on Build GRCh38 |
Assay Gene Location: | Within Intron 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEBL | NM_001173484.1 | NP_001166955.1 | ||
| NM_006393.2 | NP_006384.1 | |||
| NM_213569.2 | NP_998734.1 | |||
| XM_005252342.4 | XP_005252399.1 | |||
| XM_005252343.4 | XP_005252400.1 | |||
| XM_005252344.4 | XP_005252401.1 | |||
| XM_011519291.2 | XP_011517593.1 | |||
| XM_017015467.1 | XP_016870956.1 | |||
| XM_017015468.1 | XP_016870957.1 | |||
| AB209311.1 | BAD92548.1 | |||
| AJ580772.1 | CAE45323.1 | |||
| AK096540.1 | BAC04812.1 | |||
| AK294926.1 | ||||
| AK295186.1 | ||||
| AK316521.1 | ||||
| BC110452.1 | AAI10453.1 | |||
| BC110453.1 | AAI10454.1 | |||
| BC126132.1 | ||||
| BC126134.1 | ||||
| Y16241.1 | CAA76130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv472153 | Chr.10:20790110 - 20790110 on Build GRCh38 | Novel Sequence Insertion |
|
More Information
Set Membership: |
|

Genomic Map