Assay Details
Target Gene Details
Entrez Gene ID: | 100507633 |
Gene Name: | TNKS2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.10:91782839-91798291 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNKS2-AS1 | NR_108070.1 | 1 | 130 | |
| NR_108071.1 | ||||
| AA776692.1 | ||||
| BM678746.1 | 1 | 130 |
More Information
Additional Information:
For this assay, SNP(s) [rs73325969] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map