Assay Details
Target Gene Details
Entrez Gene ID: | 3698 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain 2 |
Gene Aliases: |
H2P, SHAP |
Location: |
Chr.10:7703273-7749520 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH2 | NM_002216.2 | NP_002207.2 | ||
| AK225277.1 | ||||
| AK225329.1 | ||||
| AK291379.1 | ||||
| AK297334.1 | ||||
| BC132685.1 | ||||
| BG617651.1 | ||||
| X07173.1 | CAA30160.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549933 | Chr.10:7586456 - 8160993 on Build GRCh38 | Gain |
|
| esv3891752 | Chr.10:7697592 - 7743053 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map