Assay Details
Target Gene Details
Entrez Gene ID: | 6865 |
Gene Name: | tachykinin receptor 2 |
Gene Aliases: |
NK2R, NKNAR, SKR, TAC2R |
Location: |
Chr.10:69404202-69416918 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TACR2 | NM_001057.2 | 1 | 81 | NP_001048.2 |
| AK291996.1 | 1 | 81 | ||
| AK300649.1 | ||||
| DA929480.1 | 1 | 81 |
More Information
Set Membership: |
|

Genomic Map