Assay Details
Target Gene Details
Entrez Gene ID: | 159686 |
Gene Name: | cilia and flagella associated protein 58 |
Gene Aliases: |
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 |
Location: |
Chr.10:104339124-104455090 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP58 | XM_011539377.2 | XP_011537679.1 | ||
| AK296463.1 |
More Information
Set Membership: |
|

Genomic Map