Assay Details
Target Gene Details
Entrez Gene ID: | 5950 |
Gene Name: | retinol binding protein 4 |
Gene Aliases: |
MCOPCB10, RDCCAS |
Location: |
Chr.10:93591836-93601744 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP4 | NM_001323517.1 | NP_001310446.1 | ||
| NM_001323518.1 | NP_001310447.1 | |||
| NM_006744.3 | NP_006735.2 | |||
| BC020633.1 | AAH20633.1 | |||
| BG565176.1 | ||||
| BI712834.1 | ||||
| BX495987.1 | ||||
| X00129.1 | CAA24959.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77196820] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map