Assay Details
Target Gene Details
Entrez Gene ID: | 55691 |
Gene Name: | FERM domain containing 4A |
Gene Aliases: |
CCAFCA, FRMD4, bA295P9.4 |
Location: |
Chr.10:13643706-14330952 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRMD4A | NM_018027.4 | NP_060497.3 | ||
| NR_134578.1 | ||||
| XM_011519543.2 | XP_011517845.1 | |||
| XM_017016394.1 | XP_016871883.1 | |||
| XM_017016395.1 | XP_016871884.1 | |||
| AB037715.1 | ||||
| AK289693.1 | ||||
| BC151244.1 |
Target Gene Details
Entrez Gene ID: | 101928453 |
Gene Name: | uncharacterized LOC101928453 |
Gene Aliases: |
- |
Location: |
Chr.10:14074284-14087881 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928453 | NR_120638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528027 | Chr.10:13988319 - 14253302 on Build GRCh38 | Gain |
|
| nsv549996 | Chr.10:13964391 - 14242607 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map