Assay Details
Target Gene Details
Entrez Gene ID: | 159686 |
Gene Name: | cilia and flagella associated protein 58 |
Gene Aliases: |
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 |
Location: |
Chr.10:104339124-104455090 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP58 | XM_011539377.2 | XP_011537679.1 | ||
| XM_017015787.1 | XP_016871276.1 | |||
| XM_017015788.1 | XP_016871277.1 | |||
| AK296463.1 |
Target Gene Details
Entrez Gene ID: | 100505869 |
Gene Name: | CFAP58 antisense RNA 1 (head to head) |
Gene Aliases: |
CCDC147-AS1 |
Location: |
Chr.10:104351301-104353575 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP58-AS1 | NR_108036.1 | |||
| BX093559.1 |
More Information
Set Membership: |
|

Genomic Map