Assay Details
Target Gene Details
Entrez Gene ID: | 55301 |
Gene Name: | oleoyl-ACP hydrolase |
Gene Aliases: |
AURA1, SAST, THEDC1 |
Location: |
Chr.10:15043896-15073852 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLAH | NM_001039702.2 | NP_001034791.1 | ||
| NM_018324.2 | NP_060794.1 | |||
| XM_017016376.1 | XP_016871865.1 | |||
| AK001844.1 | BAA91937.1 | |||
| AK001968.1 | BAA92007.1 | |||
| BC050372.1 | AAH50372.1 | |||
| BG722790.1 | ||||
| BI830605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831794 | Chr.10:14993658 - 15148891 on Build GRCh38 | Loss |
|
| nsv831793 | Chr.10:14870604 - 15058763 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map