Assay Details
Target Gene Details
Entrez Gene ID: | 83641 |
Gene Name: | family with sequence similarity 107 member B |
Gene Aliases: |
C10orf45 |
Location: |
Chr.10:14518557-14774897 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831791 | Chr.10:14505865 - 14682240 on Build GRCh38 | Gain |
|
| esv34151 | Chr.10:14344296 - 14751833 on Build GRCh38 | Loss |
|
| nsv5877 | Chr.10:14521492 - 14555125 on Build GRCh38 | Insertion |
|
| nsv528028 | Chr.10:14400348 - 14563663 on Build GRCh38 | Gain |
|
| nsv549997 | Chr.10:14459237 - 14814109 on Build GRCh38 | Gain |
|
| nsv437659 | Chr.10:14521098 - 14560444 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map