Assay Details
Target Gene Details
Entrez Gene ID: | 59 |
Gene Name: | actin, alpha 2, smooth muscle, aorta |
Gene Aliases: |
AAT6, ACTSA, MYMY5 |
Location: |
Chr.10:88935074-88991397 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTA2 | NM_001141945.2 | NP_001135417.1 | ||
| NM_001320855.1 | NP_001307784.1 | |||
| AY692464.1 | ||||
| BP248338.1 | ||||
| DC304647.1 | ||||
| DC354672.1 | ||||
| DC359878.1 | ||||
| DC385559.1 |
Target Gene Details
Entrez Gene ID: | 355 |
Gene Name: | Fas cell surface death receptor |
Gene Aliases: |
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6 |
Location: |
Chr.10:88968429-89017061 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAS | XM_006717819.3 | XP_006717882.1 | ||
| XM_011539764.2 | XP_011538066.1 | |||
| XM_011539765.2 | XP_011538067.1 | |||
| XM_011539766.2 | XP_011538068.1 | |||
| AB209361.1 | BAD92598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831943 | Chr.10:88905079 - 89106077 on Build GRCh38 | Gain |
|
| esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map