Assay Details
Target Gene Details
Entrez Gene ID: | 102724316 |
Gene Name: | SVIL antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:29409534-29487856 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SVIL-AS1 | NR_110920.1 | |||
| NR_110921.1 | ||||
| NR_110922.1 | ||||
| NR_110925.1 | ||||
| NR_110926.1 | ||||
| NR_110927.1 | ||||
| NR_110928.1 | ||||
| NR_110929.1 | ||||
| BF511092.1 | ||||
| BF739922.1 | ||||
| BM925434.1 | ||||
| BX461223.2 | ||||
| DA715011.1 | ||||
| DA952579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038080 | Chr.10:29361590 - 29565510 on Build GRCh38 | Gain |
|
| esv4280 | Chr.10:29423193 - 29423864 on Build GRCh38 | Loss |
|
| esv2518867 | Chr.10:29422808 - 29424200 on Build GRCh38 | Deletion |
|
| nsv512155 | Chr.10:29422644 - 29424461 on Build GRCh38 | Loss |
|
| nsv519798 | Chr.10:29332490 - 29500165 on Build GRCh38 | Loss |
|
| nsv1047143 | Chr.10:29108926 - 29425407 on Build GRCh38 | Gain |
|
| esv2127127 | Chr.10:29423028 - 29423988 on Build GRCh38 | Deletion |
|
| esv2734606 | Chr.10:27447765 - 29432048 on Build GRCh38 | Deletion |
|
| esv3622772 | Chr.10:29423054 - 29427511 on Build GRCh38 | Loss |
|

Genomic Map