Assay Details
Target Gene Details
Entrez Gene ID: | 159686 |
Gene Name: | cilia and flagella associated protein 58 |
Gene Aliases: |
C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1 |
Location: |
Chr.10:104339124-104455090 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP58 | XM_011539377.2 | XP_011537679.1 | ||
| XM_017015787.1 | XP_016871276.1 | |||
| AK296463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2674572 | Chr.10:104344565 - 104348112 on Build GRCh38 | Deletion |
|
| esv3624428 | Chr.10:104344569 - 104348113 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map