Assay Details
Target Gene Details
Entrez Gene ID: | 142940 |
Gene Name: | TruB pseudouridine synthase family member 1 |
Gene Aliases: |
PUS4 |
Location: |
Chr.10:114938193-114977680 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRUB1 | NM_139169.4 | NP_631908.1 | ||
| AF448144.1 | AAL40350.1 | |||
| AK223567.1 | BAD97287.1 | |||
| AK302800.1 | ||||
| AK312805.1 | ||||
| BC030601.2 | AAH30601.1 | |||
| BX648479.1 | ||||
| BX648709.1 | CAH10560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038050 | Chr.10:114555350 - 115135954 on Build GRCh38 | Gain |
|
| esv3624624 | Chr.10:114876531 - 115123608 on Build GRCh38 | Gain |
|
| esv3624625 | Chr.10:114913721 - 114991007 on Build GRCh38 | Gain |
|
| esv3577749 | Chr.10:114884410 - 115119719 on Build GRCh38 | Gain |
|
| dgv977n100 | Chr.10:114883336 - 115119719 on Build GRCh38 | Gain |
|
| nsv831997 | Chr.10:114929793 - 115095941 on Build GRCh38 | Gain |
|
| esv3624626 | Chr.10:114923110 - 115094807 on Build GRCh38 | Gain |
|
| nsv831996 | Chr.10:114841793 - 115013209 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79274267] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map