Assay Details
Target Gene Details
Entrez Gene ID: | 10276 |
Gene Name: | neuroepithelial cell transforming 1 |
Gene Aliases: |
ARHGEF8, NET1A |
Location: |
Chr.10:5412551-5459056 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NET1 | NM_001047160.2 | NP_001040625.1 | ||
| NM_005863.4 | NP_005854.2 | |||
| NR_073040.1 | ||||
| AB209852.1 | BAD93089.1 | |||
| AJ010046.1 | CAA08974.1 | |||
| AK024919.1 | ||||
| AK294244.1 | ||||
| AK299335.1 | ||||
| AK302687.1 | ||||
| AK304028.1 | ||||
| AK315566.1 | ||||
| BC010285.1 | AAH10285.1 | |||
| BC053553.1 | AAH53553.1 | |||
| BP354878.1 | ||||
| BX537509.1 | 1 | 300 | ||
| DA727151.1 | ||||
| DB177813.1 | ||||
| DC307907.1 | ||||
| S82401.1 | AAB37683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831774 | Chr.10:5421593 - 5615458 on Build GRCh38 | Gain |
|
| nsv549886 | Chr.10:5163474 - 5590097 on Build GRCh38 | Loss |
|
| nsv1045141 | Chr.10:5384177 - 5449996 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map