Assay Details
Target Gene Details
Entrez Gene ID: | 85450 |
Gene Name: | inositol 1,4,5-trisphosphate receptor interacting protein |
Gene Aliases: |
DANGER, KIAA1754, bA127L20, bA127L20.2 |
Location: |
Chr.10:104309696-104338493 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPRIP | NM_001272012.1 | 2 | 5110 | NP_001258941.1 |
| NM_001272013.1 | 2 | 5230 | NP_001258942.1 | |
| NM_033397.3 | 3 | 5422 | NP_203755.1 | |
| XM_005270257.2 | 1 | 4986 | XP_005270314.1 | |
| DA832442.1 | 1 | 312 | ||
| EL593529.1 | 1 | 295 |

Genomic Map