Assay Details
Target Gene Details
Entrez Gene ID: | 1488 |
Gene Name: | C-terminal binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.10:124987849-125161061 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2773 | Chr.10:124993894 - 124998042 on Build GRCh38 | Loss |
|
| dgv51n21 | Chr.10:124995565 - 125004911 on Build GRCh38 | Loss |
|
| nsv518907 | Chr.10:124993119 - 125002611 on Build GRCh38 | Loss |
|
| nsv552289 | Chr.10:124875780 - 124998761 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72828985] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map