Assay Details
Target Gene Details
Entrez Gene ID: | 5728 |
Gene Name: | phosphatase and tensin homolog |
Gene Aliases: |
10q23del, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, PTEN1, TEP1 |
Location: |
Chr.10:87863438-87971930 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTEN | NM_000314.6 | NP_000305.3 | ||
| NM_001304717.2 | NP_001291646.2 | |||
| NM_001304718.1 | NP_001291647.1 | |||
| AK313581.1 | ||||
| BC005821.2 | AAH05821.1 | |||
| CR450306.1 | CAG29302.1 | |||
| JF268690.1 | ||||
| KJ534922.1 | ||||
| U92436.1 | AAC51182.1 | |||
| U93051.1 | AAC51183.1 | |||
| U96180.1 | AAB66902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551832 | Chr.10:87870667 - 87891002 on Build GRCh38 | Loss |
|
| nsv1050406 | Chr.10:87872381 - 87898149 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map