Assay Details
Target Gene Details
Entrez Gene ID: | 3185 |
Gene Name: | heterogeneous nuclear ribonucleoprotein F |
Gene Aliases: |
HNRPF, OK/SW-cl.23, mcs94-1 |
Location: |
Chr.10:43385617-43409248 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPF | NM_001098204.1 | NP_001091674.1 | ||
| NM_001098205.1 | NP_001091675.1 | |||
| NM_001098206.1 | NP_001091676.1 | |||
| NM_001098207.1 | NP_001091677.1 | |||
| NM_001098208.1 | NP_001091678.1 | |||
| NM_004966.3 | NP_004957.1 | |||
| AK001364.1 | ||||
| AK296696.1 | ||||
| AL542769.3 | ||||
| BC001432.2 | AAH01432.1 | |||
| BC004254.1 | AAH04254.1 | |||
| BC015580.2 | AAH15580.1 | |||
| BC016736.1 | AAH16736.1 | |||
| BC106008.1 | AAI06009.1 | |||
| CR976524.1 | ||||
| DA535143.1 | ||||
| DA594075.1 | ||||
| DA679417.1 | ||||
| DB011453.1 | ||||
| DB049909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv146n67 | Chr.10:43387970 - 43454488 on Build GRCh38 | Gain |
|
| nsv831842 | Chr.10:43351093 - 43492053 on Build GRCh38 | Loss |
|
| esv3623043 | Chr.10:43391117 - 43392506 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map