Assay Details
Target Gene Details
Entrez Gene ID: | 8028 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10 |
Gene Aliases: |
AF10 |
Location: |
Chr.10:21533349-21743630 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLLT10 | NM_001195626.1 | NP_001182555.1 | ||
| NM_001324297.1 | NP_001311226.1 | |||
| NM_004641.3 | NP_004632.1 | |||
| NR_136736.1 | ||||
| AB209755.1 | BAD92992.1 | |||
| AY598745.1 | AAT47519.1 | |||
| U13948.1 | AAA79972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv550202 | Chr.10:21567546 - 21738715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map