Assay Details
Target Gene Details
Entrez Gene ID: | 7580 |
Gene Name: | zinc finger protein 32 |
Gene Aliases: |
KOX30 |
Location: |
Chr.10:43643859-43649350 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF32 | NM_001005368.2 | NP_001005368.1 | ||
| NM_001324164.1 | NP_001311093.1 | |||
| NM_001324165.1 | NP_001311094.1 | |||
| NM_001324166.1 | NP_001311095.1 | |||
| NM_001324167.1 | NP_001311096.1 | |||
| NM_001324168.1 | NP_001311097.1 | |||
| NM_006973.2 | NP_008904.1 | |||
| XM_017016616.1 | 1 | 1508 | XP_016872105.1 | |
| AI871265.1 | ||||
| AK309483.1 | ||||
| AK312132.1 | ||||
| BG477527.1 | ||||
| HY045180.1 | ||||
| HY148951.1 | ||||
| U69645.1 | AAB09561.1 |
Target Gene Details
Entrez Gene ID: | 414208 |
Gene Name: | ZNF32 antisense RNA 2 |
Gene Aliases: |
ZNF32OS2, b402L1.7 |
Location: |
Chr.10:43645942-43648019 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF32-AS2 | NR_047558.1 | 3 | 729 | |
| AW188264.1 | 1 | 179 |
Target Gene Details
Entrez Gene ID: | 414201 |
Gene Name: | ZNF32 antisense RNA 3 |
Gene Aliases: |
C10orf44, ZNF32-OT1, bA402L1.3 |
Location: |
Chr.10:43628817-43674699 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF32-AS3 | NR_038867.1 |
More Information
Set Membership: |
|

Genomic Map