Assay Details
Target Gene Details
Entrez Gene ID: | 8028 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10 |
Gene Aliases: |
AF10 |
Location: |
Chr.10:21533349-21743630 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLLT10 | NM_001195626.1 | NP_001182555.1 | ||
| NM_001195627.1 | NP_001182556.1 | |||
| NM_001195628.1 | NP_001182557.1 | |||
| NM_001195630.1 | NP_001182559.1 | |||
| NM_001324296.1 | NP_001311225.1 | |||
| NM_001324297.1 | NP_001311226.1 | |||
| NM_004641.3 | NP_004632.1 | |||
| NR_136736.1 | ||||
| AB209755.1 | BAD92992.1 | |||
| AF060934.1 | AAC16706.1 | |||
| AF060935.1 | AAC16707.1 | |||
| AF060936.1 | AAC16708.1 | |||
| AF060937.1 | AAC16709.1 | |||
| AF060938.1 | AAC16710.1 | |||
| AY598745.1 | AAT47519.1 | |||
| BC080577.1 | AAH80577.1 | |||
| U13948.1 | AAA79972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052477 | Chr.10:21521341 - 21600797 on Build GRCh38 | Gain |
|
| dgv133n67 | Chr.10:21450598 - 21585405 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map