Assay Details
Target Gene Details
Entrez Gene ID: | 56889 |
Gene Name: | transmembrane 9 superfamily member 3 |
Gene Aliases: |
EP70-P-iso, SMBP |
Location: |
Chr.10:96518110-96587052 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM9SF3 | NM_020123.3 | 15 | 5726 | NP_064508.3 |
| XM_011539977.1 | 15 | 5855 | XP_011538279.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577627 | Chr.10:96515251 - 96710544 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map