Assay Details
Target Gene Details
Entrez Gene ID: | 256536 |
Gene Name: | transcription elongation regulator 1 like |
Gene Aliases: |
- |
Location: |
Chr.10:131092391-131311721 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCERG1L | NM_174937.3 | NP_777597.2 | ||
| XM_011539577.1 | XP_011537879.1 | |||
| AK096269.1 | ||||
| BC042951.1 | AAH42951.1 | |||
| BC093639.1 | AAH93639.1 | |||
| BC101535.1 | AAI01536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832027 | Chr.10:131056320 - 131214052 on Build GRCh38 | Gain |
|
| nsv1042634 | Chr.10:131116646 - 131247825 on Build GRCh38 | Loss |
|
| nsv1048253 | Chr.10:129847356 - 131188858 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| nsv470974 | Chr.10:131049527 - 131126683 on Build GRCh38 | Gain |
|
| nsv428242 | Chr.10:130607951 - 131583141 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74160848] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map