Assay Details
Target Gene Details
Entrez Gene ID: | 100507143 |
Gene Name: | long intergenic non-protein coding RNA 708 |
Gene Aliases: |
GS1-756B1.2 |
Location: |
Chr.10:8259331-8268305 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00708 | NR_108058.1 |

Genomic Map