Assay Details
Target Gene Details
Entrez Gene ID: | 10539 |
Gene Name: | glutaredoxin 3 |
Gene Aliases: |
GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3 |
Location: |
Chr.10:130136375-130182877 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLRX3 | NM_001199868.1 | NP_001186797.1 | ||
| NM_001321980.1 | NP_001308909.1 | |||
| NM_006541.4 | NP_006532.2 | |||
| XM_017015474.1 | XP_016870963.1 | |||
| AF118649.1 | AAF28841.1 | |||
| AF118652.1 | AAF28844.1 | |||
| AJ010841.1 | CAA09375.1 | |||
| AK022131.1 | ||||
| AK225609.1 | ||||
| BC005289.1 | AAH05289.1 | |||
| BP347054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048253 | Chr.10:129847356 - 131188858 on Build GRCh38 | Gain |
|
| nsv832024 | Chr.10:129947474 - 130147836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map