Assay Details
Target Gene Details
Entrez Gene ID: | 51207 |
Gene Name: | dual specificity phosphatase 13 |
Gene Aliases: |
BEDP, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP |
Location: |
Chr.10:75094432-75109221 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516793 | Chr.10:75094806 - 75108141 on Build GRCh38 | Gain |
|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
| nsv7199 | Chr.10:75077582 - 75112320 on Build GRCh38 | Insertion |
|
| nsv518639 | Chr.10:75083167 - 75108141 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map