Assay Details
Target Gene Details
Entrez Gene ID: | 79741 |
Gene Name: | coiled-coil domain containing 7 |
Gene Aliases: |
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68 |
Location: |
Chr.10:32446082-32882864 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |

Genomic Map