Assay Details
Target Gene Details
Entrez Gene ID: | 219670 |
Gene Name: | enkurin, TRPC channel interacting protein |
Gene Aliases: |
C10orf63, CFAP106 |
Location: |
Chr.10:24981979-25062279 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENKUR | NM_001270383.1 | NP_001257312.1 | ||
| NM_145010.3 | NP_659447.1 | |||
| NR_072992.1 | ||||
| NR_072993.1 | ||||
| XM_006717389.1 | XP_006717452.1 | |||
| XM_006717390.1 | XP_006717453.1 | |||
| XM_011519348.1 | XP_011517650.1 | |||
| XM_017015830.1 | XP_016871319.1 | |||
| AK095021.1 | BAC04477.1 | |||
| AK292495.1 | ||||
| AY454125.1 | AAS45168.1 | |||
| BC026165.2 | AAH26165.1 | |||
| CR936720.1 | ||||
| DA285403.1 |
Target Gene Details
Entrez Gene ID: | 79896 |
Gene Name: | threonine synthase like 1 |
Gene Aliases: |
TSH1 |
Location: |
Chr.10:24952319-25026664 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THNSL1 | XM_017016665.1 | XP_016872154.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77260255] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map