Assay Details
Target Gene Details
Entrez Gene ID: | 8861 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM-2, CLIM2, LDB-1, NLI |
Location: |
Chr.10:102102100-102120473 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDB1 | XM_017016867.1 | XP_016872356.1 | ||
| XM_017016868.1 | XP_016872357.1 |
More Information
Set Membership: |
|

Genomic Map